siRNA Details
Gene Symbol: Cldnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028849.1 | NP_083125.1 | 3 | 349 | ||
GenBank mRNA | AK006929.1 | - | 3 | 384 | ||
BC128277.1 | - | 3 | 349 | 40091490 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cldnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028849.1 | NP_083125.1 | 3 | 349 | ||
GenBank mRNA | AK006929.1 | - | 3 | 384 | ||
BC128277.1 | - | 3 | 349 | 40091490 |