siRNA Details
| Gene Symbol: Mtmr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028860.2 | NP_083136.2 | 7 | 802 | ||
| GenBank mRNA | AK141138.1 | - | 7 | 782 | ||
| BC032166.1 | - | 7 | 785 | 5352919 | ||
| BC081544.1 | - | 7 | 786 | 3672099 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr3| Gene Symbol: Mtmr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028860.2 | NP_083136.2 | 7 | 802 | ||
| GenBank mRNA | AK141138.1 | - | 7 | 782 | ||
| BC032166.1 | - | 7 | 785 | 5352919 | ||
| BC081544.1 | - | 7 | 786 | 3672099 | ||