siRNA Details
| Gene Symbol: Lrrtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028880.3 | NP_083156.2 | 2 | 778 | ||
| GenBank mRNA | AK014545.1 | - | 2 | 781 | ||
| AK053345.1 | - | 2 | 768 | |||
| AK053364.1 | - | 2 | 402 | |||
| AK080836.1 | - | 2 | 809 | |||
| AK136857.1 | - | 1 | 277 | |||
| AY182025.1 | - | 1 | 213 | |||
| BC027803.1 | - | 2 | 660 | 5321979 | ||

Lrrtm1
Genomic Map