siRNA Details
| Gene Symbol: Crtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028881.2 | NP_083157.1 | 13 | 1886 | ||
| GenBank mRNA | AA276938.1 | - | 2 | 111 | ||
| AA611164.1 | - | 3 | 606 | |||
| AK014553.1 | - | 7 | 1993 | |||
| AK079382.1 | - | 10 | 2227 | |||
| AK156192.1 | - | 13 | 1865 | |||
| AK169944.1 | - | 9 | 1535 | |||
| BC019713.1 | - | 3 | 268 | |||
| BC023091.1 | - | 9 | 1476 | 4023064 | ||
| BC023831.1 | - | 13 | 1881 | 5345301 | ||
| BC032183.1 | - | 13 | 1891 | 5310152 | ||
| BC033300.1 | - | 13 | 1881 | 4166411 | ||

Crtc2
Genomic Map