siRNA Details
| Gene Symbol: Tldc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028883.2 | NP_083159.1 | 3 | 394 | ||
| GenBank mRNA | AK014579.1 | - | 3 | 380 | ||
| AK038974.1 | - | 3 | 375 | |||
| AK079186.1 | - | 3 | 394 | |||
| AK079239.1 | - | 3 | 394 | |||
| BC030874.1 | - | 2 | 334 | 4224306 | ||

Tldc1
Genomic Map