siRNA Details
| Gene Symbol: Zfp84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023750.2 | NP_076239.1 | 5 | 1268 | ||
| GenBank mRNA | AK014610.1 | - | 5 | 1224 | ||
| AK034259.1 | - | 5 | 1163 | |||
| AK083748.1 | - | 5 | 1209 | |||
| AK158422.1 | - | 5 | 1218 | |||
| BC008540.1 | - | 5 | 1216 | 3495627 | ||

Zfp84
Genomic Map