siRNA Details
| Gene Symbol: Smchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028887.3 | NP_083163.3 | 42 | 5472 | ||
| GenBank mRNA | AK016419.1 | - | 17 | 1949 | ||
| AK030247.1 | - | 10 | 1054 | |||
| AK129181.1 | - | 20 | 2469 | |||
| BC044905.1 | - | 6 | 619 | |||
| BC058205.1 | - | 8 | 932 | |||
| BC058618.1 | - | 18 | 2124 | |||
| BC062946.1 | - | 28 | 3317 | 30357469 | ||
| BC076627.1 | - | 35 | 4435 | |||

Smchd1
Genomic Map