siRNA Details
| Gene Symbol: 4931428F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166394.1 | NP_001159866.1 | 2 | 669 | ||
| NM_028888.2 | NP_083164.1 | 3 | 1066 | |||
| GenBank mRNA | AK016481.1 | - | 2 | 1004 | ||
| AK136929.1 | - | 2 | 831 | |||
| BC043046.1 | - | 2 | 826 | 5703896 | ||
| BC059045.1 | - | 2 | 604 | 6816353 | ||

4931428F04Rik
Genomic Map