siRNA Details
| Gene Symbol: Gcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028900.4 | NP_083176.3 | 2 | 2586 | ||
| GenBank mRNA | AK016531.1 | - | 2535 | |||
| AK032253.1 | - | 2 | 2586 | |||
| BC051389.1 | - | 1 | 160 | |||
| BC066807.1 | - | 2 | 2555 | 30101518 | ||

Gcc1
Genomic Map