siRNA Details
| Gene Symbol: Gcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028900.4 | NP_083176.3 | 2 | 1887 | ||
| GenBank mRNA | AK016531.1 | - | 1836 | |||
| AK032253.1 | - | 2 | 1887 | |||
| BC066807.1 | - | 2 | 1856 | 30101518 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gcc1| Gene Symbol: Gcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028900.4 | NP_083176.3 | 2 | 1887 | ||
| GenBank mRNA | AK016531.1 | - | 1836 | |||
| AK032253.1 | - | 2 | 1887 | |||
| BC066807.1 | - | 2 | 1856 | 30101518 | ||