siRNA Details
| Gene Symbol: Myo18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028901.2 | NP_083177.2 | 14 | 3096 | ||
| GenBank mRNA | AK016515.1 | - | 9 | 1224 | ||
| BC145218.1 | - | 8 | 1045 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo18b| Gene Symbol: Myo18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028901.2 | NP_083177.2 | 14 | 3096 | ||
| GenBank mRNA | AK016515.1 | - | 9 | 1224 | ||
| BC145218.1 | - | 8 | 1045 | |||