siRNA Details
| Gene Symbol: 4932438H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163695.1 | NP_001157167.1 | 2 | 484 | ||
| NM_028905.3 | NP_083181.1 | 3 | 732 | |||
| GenBank mRNA | AK016554.1 | - | 3 | 733 | ||
| AK035767.1 | - | 2 | 474 | |||
| AK043035.1 | - | 3 | 722 | |||
| BC032975.1 | - | 1 | 631 | 1264334 | ||

4932438H23Rik
Genomic Map