siRNA Details
| Gene Symbol: Dpp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028906.2 | NP_083182.2 | 14 | 1922 | ||
| GenBank mRNA | AK016546.1 | - | 14 | 1961 | ||
| AK029788.1 | - | 12 | 3793 | |||
| AK155605.1 | - | 14 | 1967 | |||
| BC043124.1 | - | 14 | 1922 | 6410075 | ||
| BC059222.1 | - | 14 | 1922 | 6410075 | ||

Dpp8
Genomic Map