siRNA Details
| Gene Symbol: Efhc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028916.4 | NP_083192.2 | 11, 12 | 1841 | ||
| GenBank mRNA | AK016714.1 | - | 11, 12 | 1800 | ||
| AK029501.1 | - | 11, 12 | 1807 | |||
| BC120659.1 | - | 11, 12 | 1776 | 40129582 | ||
| BC125625.1 | - | 11, 12 | 1776 | 40130140 | ||

Efhc2
Genomic Map