siRNA Details
| Gene Symbol: Ttll11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029774.2 | NP_084050.1 | 5 | 1339 | ||
| GenBank mRNA | AK016577.1 | - | 5 | 796 | ||
| AM690755.1 | - | 5 | 1339 | |||
| BC115858.1 | - | 3 | 788 | 40057265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttll11| Gene Symbol: Ttll11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029774.2 | NP_084050.1 | 5 | 1339 | ||
| GenBank mRNA | AK016577.1 | - | 5 | 796 | ||
| AM690755.1 | - | 5 | 1339 | |||
| BC115858.1 | - | 3 | 788 | 40057265 | ||