siRNA Details
| Gene Symbol: Ttll11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029774.2 | NP_084050.1 | 4 | 772 | ||
| GenBank mRNA | AM690755.1 | - | 4 | 772 | ||
| BC115858.1 | - | 2 | 221 | 40057265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttll11| Gene Symbol: Ttll11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029774.2 | NP_084050.1 | 4 | 772 | ||
| GenBank mRNA | AM690755.1 | - | 4 | 772 | ||
| BC115858.1 | - | 2 | 221 | 40057265 | ||