siRNA Details
| Gene Symbol: Gle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028923.3 | NP_083199.1 | 10 | 1496 | ||
| GenBank mRNA | AK016671.1 | - | 10 | 1478 | ||
| AK028183.1 | - | 10 | 1480 | |||
| AK133888.1 | - | 10 | 1448 | |||
| AK145748.1 | - | 10 | 1472 | |||
| AK161015.1 | - | 10 | 1442 | |||
| AK161659.1 | - | 10 | 1479 | |||
| AK183599.1 | - | 4 | 327 | |||
| BC026797.1 | - | 10 | 1476 | 3990668 | ||

Gle1
Genomic Map