siRNA Details
| Gene Symbol: Gle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028923.3 | NP_083199.1 | 2 | 252 | ||
| GenBank mRNA | AK016671.1 | - | 2 | 234 | ||
| AK028183.1 | - | 2 | 236 | |||
| AK029395.1 | - | 2 | 206 | |||
| AK133888.1 | - | 2 | 204 | |||
| AK145748.1 | - | 2 | 228 | |||
| AK150230.1 | - | 2 | 227 | |||
| AK161015.1 | - | 2 | 198 | |||
| AK161659.1 | - | 2 | 235 | |||
| BC026797.1 | - | 2 | 235 | 3990668 | ||

Gle1
Genomic Map