siRNA Details
| Gene Symbol: Gle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028923.3 | NP_083199.1 | 12 | 1786 | ||
| GenBank mRNA | AK016671.1 | - | 12 | 1768 | ||
| AK028183.1 | - | 12 | 1770 | |||
| AK133888.1 | - | 12 | 1738 | |||
| AK145748.1 | - | 12 | 1762 | |||
| AK161015.1 | - | 12 | 1732 | |||
| AK161659.1 | - | 12 | 1769 | |||
| AK180772.1 | - | 2 | 218 | |||
| AK207245.1 | - | 2 | 200 | |||
| AK210720.1 | - | 2 | 200 | |||
| AK216189.1 | - | 2 | 200 | |||
| BC026797.1 | - | 12 | 1766 | 3990668 | ||

Gle1
Genomic Map