siRNA Details
| Gene Symbol: Lrriq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028938.2 | NP_083214.2 | 4 | 925 | ||
| GenBank mRNA | AK015333.1 | - | 4 | 822 | ||
| AK015758.1 | - | 4 | 772 | |||
| AK016634.1 | - | 4 | 820 | |||
| AK132656.1 | - | 4 | 890 | |||
| BC112424.1 | - | 4 | 782 | 40062088 | ||
| BC113130.1 | - | 4 | 782 | 40062093 | ||

Lrriq3
Genomic Map