siRNA Details
| Gene Symbol: 4933402E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199996.1 | NP_001186925.1 | 4 | 872 | ||
| NM_001199997.1 | NP_001186926.1 | 4 | 833 | |||
| GenBank mRNA | AK016614.1 | - | 4 | 875 | ||
| BC132156.1 | - | 4 | 868 | 40130433 | ||

4933402E13Rik
Genomic Map