siRNA Details
| Gene Symbol: Cmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163262.1 | NP_001156734.1 | 10 | 1143 | ||
| NM_028941.1 | NP_083217.1 | 10 | 966 | |||
| GenBank mRNA | AK016712.1 | - | 10 | 967 | ||
| AK085367.1 | - | 10 | 1046 | |||
| AK173234.1 | - | 4 | 3393 | |||
| BC028834.1 | - | 8 | 635 | |||
| BC158118.1 | - | 11 | 1020 | 9088191 | ||
| CA552751.1 | - | 7 | 534 | |||

Cmip
Genomic Map