siRNA Details
| Gene Symbol: Cmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163262.1 | NP_001156734.1 | 7 | 855 | ||
| NM_028941.1 | NP_083217.1 | 7 | 678 | |||
| GenBank mRNA | AK016712.1 | - | 7 | 679 | ||
| AK085367.1 | - | 7 | 758 | |||
| AK173234.1 | - | 1 | 3105 | |||
| BC028834.1 | - | 5 | 347 | |||
| BC158118.1 | - | 8 | 732 | 9088191 | ||
| CA552751.1 | - | 4 | 246 | |||

Cmip
Genomic Map