siRNA Details
| Gene Symbol: Slco6c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028942.4 | NP_083218.1 | 7 | 1266 | ||
| GenBank mRNA | AK016647.1 | - | 7 | 1214 | ||
| AK029505.1 | - | 7 | 1267 | |||
| AK132668.1 | - | 7 | 1217 | |||
| AK161249.1 | - | 7 | 1217 | |||
| BC139066.1 | - | 7 | 1169 | 8862088 | ||
| BC139067.1 | - | 7 | 1169 | 8862089 | ||

Slco6c1
Genomic Map