siRNA Details
| Gene Symbol: Slc9b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028946.3 | NP_083222.2 | 6 | 1159 | ||
| GenBank mRNA | AK016883.1 | - | 6 | 1011 | ||
| AK016917.1 | - | 6 | 1139 | |||
| AK029525.1 | - | 6 | 1160 | |||
| BC025002.1 | - | 5 | 1081 | |||
| EU846100.1 | - | 5 | 690 | |||

Slc9b1
Genomic Map