siRNA Details
| Gene Symbol: Cfap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028948.2 | NP_083224.2 | 3 | 547 | ||
| GenBank mRNA | AK016821.1 | - | 4 | 758 | ||
| BC099500.1 | - | 3 | 465 | 30932292 | ||
| BC125572.1 | - | 3 | 437 | 40130087 | ||
| BC125574.1 | - | 3 | 437 | 40130089 | ||

Cfap53
Genomic Map