siRNA Details
| Gene Symbol: Zswim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029912.2 | NP_001025083.1 | 13 | 2745 | ||
| GenBank mRNA | AK122519.1 | - | 13 | 2745 | ||
| BC089375.1 | - | 2 | 159 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zswim5| Gene Symbol: Zswim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029912.2 | NP_001025083.1 | 13 | 2745 | ||
| GenBank mRNA | AK122519.1 | - | 13 | 2745 | ||
| BC089375.1 | - | 2 | 159 | |||