siRNA Details
| Gene Symbol: Zswim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029912.2 | NP_001025083.1 | 7 | 1723 | ||
| GenBank mRNA | AK122519.1 | - | 7 | 1723 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zswim5| Gene Symbol: Zswim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029912.2 | NP_001025083.1 | 7 | 1723 | ||
| GenBank mRNA | AK122519.1 | - | 7 | 1723 | ||