siRNA Details
| Gene Symbol: Pus10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033654.2 | NP_001028826.1 | 8, 9 | 799 | ||
| NM_028304.2 | NP_082580.1 | 8, 9 | 1212 | |||
| NM_028956.4 | NP_083232.1 | 8, 9 | 809 | |||
| GenBank mRNA | AK012737.1 | - | 8, 9 | 976 | ||
| AK017063.1 | - | 8, 9 | 786 | |||
| AK028368.1 | - | 8, 9 | 796 | |||
| AK029866.1 | - | 8, 9 | 800 | |||
| AK088562.1 | - | 8, 9 | 800 | |||
| AK145055.1 | - | 8, 9 | 1158 | |||
| BC025555.1 | - | 8, 9 | 1030 | 5321811 | ||

Pus10
Genomic Map