siRNA Details
| Gene Symbol: CEACAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002483.6 | NP_002474.4 | 4 | 1000 | ||
| XM_011526990.1 | XP_011525292.1 | 4 | 1099 | |||
| GenBank mRNA | AK312542.1 | - | 4 | 985 | ||
| BC005008.1 | - | 4 | 921 | 3640231 | ||
| BT009774.1 | - | 4 | 881 | |||
| M18216.1 | - | 4 | 927 | |||
| M18728.1 | - | 4 | 931 | |||
| M29541.1 | - | 4 | 974 | |||

CEACAM6
Genomic Map