siRNA Details
| Gene Symbol: 4933416C03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161855.1 | NP_001155327.1 | 1 | 710 | ||
| GenBank mRNA | AK133409.1 | - | 1 | 673 | ||
| BC147630.1 | - | 1 | 495 | |||
| BC147644.1 | - | 1 | 495 | |||
| Gene Symbol: Taf7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028958.4 | NP_083234.2 | 5, 6 | 711 | ||
| GenBank mRNA | AF285574.1 | - | 5, 6 | 515 | ||
| AK017109.1 | - | 5, 6 | 712 | |||
| BC106854.2 | - | 5, 6 | 503 | 40046501 | ||

4933416C03Rik
Genomic Map