siRNA Details
| Gene Symbol: Snx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163389.1 | NP_001156861.1 | 7, 8 | 747 | ||
| NM_028965.4 | NP_083241.1 | 8, 9 | 807 | |||
| GenBank mRNA | AK017118.1 | - | 3, 4 | 413 | ||
| AK042409.1 | - | 7, 8 | 723 | |||
| AK076332.1 | - | 8, 9 | 783 | |||
| BC014719.1 | - | 7, 8 | 724 | 4236099 | ||

Snx11
Genomic Map