siRNA Details
Gene Symbol: Samd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037221.2 | NP_001032298.1 | 7 | 1853 | ||
NM_001163433.1 | NP_001156905.1 | 6 | 1333 | |||
NM_028966.3 | NP_083242.1 | 6 | 1589 | |||
GenBank mRNA | AK017081.1 | - | 2 | 167 | ||
AK034323.1 | - | 5 | 1183 | |||
AK133470.1 | - | 6 | 1568 | |||
AK134243.1 | - | 7 | 1851 | |||
AK161277.1 | - | 6 | 1327 | |||
AK163751.1 | - | 5 | 1610 | |||
BC049740.1 | - | 2 | 139 | |||
BC145827.1 | - | 6 | 1455 | 40131130 | ||
BC145829.1 | - | 6 | 1454 | 40131132 | ||
DQ278487.1 | - | 6 | 1366 |
