siRNA Details
| Gene Symbol: Samd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037221.2 | NP_001032298.1 | 3 | 924 | ||
| NM_001163433.1 | NP_001156905.1 | 2 | 404 | |||
| NM_028966.3 | NP_083242.1 | 3 | 924 | |||
| GenBank mRNA | AK034323.1 | - | 2 | 518 | ||
| AK133470.1 | - | 3 | 903 | |||
| AK161277.1 | - | 2 | 398 | |||
| AK163751.1 | - | 2 | 945 | |||
| BC145827.1 | - | 3 | 790 | 40131130 | ||
| BC145829.1 | - | 3 | 789 | 40131132 | ||
| DQ278487.1 | - | 3 | 701 | |||

Samd4
Genomic Map