siRNA Details
| Gene Symbol: Gorasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028976.2 | NP_083252.1 | 5 | 528 | ||
| GenBank mRNA | AK154986.1 | - | 5 | 505 | ||
| AK170627.1 | - | 5 | 492 | |||
| BC012251.1 | - | 5 | 528 | 4166619 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gorasp1| Gene Symbol: Gorasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028976.2 | NP_083252.1 | 5 | 528 | ||
| GenBank mRNA | AK154986.1 | - | 5 | 505 | ||
| AK170627.1 | - | 5 | 492 | |||
| BC012251.1 | - | 5 | 528 | 4166619 | ||