siRNA Details
| Gene Symbol: Lrrc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028977.1 | NP_083253.1 | 4 | 1505 | ||
| GenBank mRNA | AK018071.1 | - | 4 | 1471 | ||
| BC030317.1 | - | 4 | 1424 | 5372009 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc17| Gene Symbol: Lrrc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028977.1 | NP_083253.1 | 4 | 1505 | ||
| GenBank mRNA | AK018071.1 | - | 4 | 1471 | ||
| BC030317.1 | - | 4 | 1424 | 5372009 | ||