siRNA Details
| Gene Symbol: Mgme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289630.1 | NP_001276559.1 | 4, 5 | 1368 | ||
| NM_001289631.1 | NP_001276560.1 | 4, 5 | 1039 | |||
| NM_028984.5 | NP_083260.1 | 4, 5 | 1097 | |||
| GenBank mRNA | AK018387.1 | - | 4, 5 | 1026 | ||
| AK031535.1 | - | 4, 5 | 967 | |||
| AK043140.1 | - | 4, 5 | 1036 | |||
| AK049565.1 | - | 4, 5 | 1238 | |||
| AK159286.1 | - | 4, 5 | 1019 | |||
| AK167919.1 | - | 4, 5 | 1026 | |||
| BC063088.1 | - | 4, 5 | 1012 | 6329248 | ||

Mgme1
Genomic Map