siRNA Details
| Gene Symbol: Themis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028998.1 | NP_083274.1 | 2 | 267 | ||
| GenBank mRNA | AK018663.1 | - | 2 | 268 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Themis3| Gene Symbol: Themis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028998.1 | NP_083274.1 | 2 | 267 | ||
| GenBank mRNA | AK018663.1 | - | 2 | 268 | ||