siRNA Details
| Gene Symbol: Nkx6-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029002.2 | NP_083278.1 | 2 | 720 | ||
| GenBank mRNA | AK018683.1 | - | 2 | 721 | ||
| AK147163.1 | - | 2 | 692 | |||
| BC140994.1 | - | 2 | 591 | 9055606 | ||
| DQ093570.1 | - | 2 | 406 | |||

Nkx6-3
Genomic Map