siRNA Details
| Gene Symbol: Rasgef1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029004.1 | NP_083280.1 | 12 | 1406 | ||
| GenBank mRNA | AK018593.1 | - | 12 | 1395 | ||
| AK039330.1 | - | 1770 | ||||
| AK047088.1 | - | 11 | 1285 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasgef1c| Gene Symbol: Rasgef1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029004.1 | NP_083280.1 | 12 | 1406 | ||
| GenBank mRNA | AK018593.1 | - | 12 | 1395 | ||
| AK039330.1 | - | 1770 | ||||
| AK047088.1 | - | 11 | 1285 | |||