siRNA Details
| Gene Symbol: Rasgef1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029004.1 | NP_083280.1 | 2 | 307 | ||
| GenBank mRNA | AK018593.1 | - | 2 | 296 | ||
| AK039330.1 | - | 671 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasgef1c| Gene Symbol: Rasgef1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029004.1 | NP_083280.1 | 2 | 307 | ||
| GenBank mRNA | AK018593.1 | - | 2 | 296 | ||
| AK039330.1 | - | 671 | ||||