siRNA Details
| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029869.1 | NP_084145.1 | 2 | 494 | ||
| NM_133906.4 | NP_598667.2 | 2 | 500 | |||
| GenBank mRNA | AK020039.1 | - | 2 | 476 | ||
| AK028932.1 | - | 2 | 431 | |||
| AK031759.1 | - | 2 | 481 | |||
| AK036807.1 | - | 2 | 483 | |||
| AK054323.1 | - | 2 | 471 | |||
| AK131148.1 | - | 1 | 639 | |||
| AK135999.1 | - | 2 | 450 | |||
| AK154831.1 | - | 2 | 480 | |||
| BC052441.1 | - | 2 | 458 | 6406518 | ||

Zkscan1
Genomic Map