siRNA Details
Gene Symbol: Lvrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029008.1 | NP_083284.1 | 2 | 923 | ||
GenBank mRNA | AK014652.1 | - | 2 | 924 | ||
BC113158.1 | - | 2 | 914 | 40058444 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_577617.
Panther Classification:
Gene Ontology Categories:
