siRNA Details
| Gene Symbol: Cds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173370.3 | NP_775546.2 | 11, 12 | 1403 | ||
| GenBank mRNA | AF533367.2 | - | 11, 12 | 1312 | ||
| BC026505.1 | - | 6, 7 | 570 | |||
| BC043707.1 | - | 2, 3 | 144 | |||
| BC055292.1 | - | 11, 12 | 1403 | 6491596 | ||

Cds1
Genomic Map