siRNA Details
| Gene Symbol: Cd200r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128132.1 | NP_001121604.1 | 3 | 416 | ||
| NM_001128133.1 | NP_001121605.1 | 3 | 416 | |||
| NM_027578.1 | NP_081854.1 | 3 | 416 | |||
| NM_029018.4 | NP_083294.2 | 3 | 416 | |||
| GenBank mRNA | AK014637.1 | - | 3 | 390 | ||
| AK014671.1 | - | 3 | 353 | |||
| AK040868.1 | - | 3 | 403 | |||
| AK170363.1 | - | 3 | 346 | |||
| AY230199.1 | - | 3 | 390 | |||
| AY703837.1 | - | 3 | 321 | |||
| AY703838.1 | - | 3 | 321 | |||
| AY703839.1 | - | 3 | 321 | |||
| AY703840.1 | - | 3 | 321 | |||
| AY703841.1 | - | 3 | 321 | |||
| AY703842.1 | - | 3 | 321 | |||
| BC106838.2 | - | 3 | 362 | 40044906 | ||

Cd200r3
Genomic Map