siRNA Details
| Gene Symbol: Scrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029022.1 | NP_083298.1 | 5 | 644 | ||
| GenBank mRNA | AK014701.1 | - | 5 | 638 | ||
| AK028703.1 | - | 5 | 618 | |||
| AK143725.1 | - | 5 | 641 | |||
| AK165936.1 | - | 5 | 641 | |||
| BC015296.1 | - | 4 | 578 | |||
| BC139390.1 | - | 5 | 639 | 8862412 | ||
| BC139391.1 | - | 5 | 639 | 8862413 | ||

Scrn3
Genomic Map