siRNA Details
Gene Symbol: Spdyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029048.3 | NP_083324.3 | 7 | 1616 | ||
GenBank mRNA | AK015441.1 | - | 7 | 1617 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spdyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029048.3 | NP_083324.3 | 7 | 1616 | ||
GenBank mRNA | AK015441.1 | - | 7 | 1617 |