siRNA Details
| Gene Symbol: Wdr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159527.1 | NP_001152999.1 | 4 | 441 | ||
| NM_172470.3 | NP_766058.3 | 4 | 441 | |||
| GenBank mRNA | AK015490.1 | - | 4 | 406 | ||
| AK029896.1 | - | 4 | 396 | |||
| AK031061.1 | - | 4 | 432 | |||
| AK053724.1 | - | 4 | 396 | |||
| AK083971.1 | - | 4 | 386 | |||
| AK173151.1 | - | 4 | 383 | |||
| BC056925.1 | - | 4 | 550 | 5704780 | ||

Wdr35
Genomic Map