siRNA Details
| Gene Symbol: Slc25a54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029054.1 | NP_083330.1 | 4 | 457 | ||
| GenBank mRNA | AK015371.1 | - | 4 | 488 | ||
| BC147589.1 | - | 4 | 457 | 9056579 | ||
| BC147621.1 | - | 4 | 457 | 9056611 | ||
| BC172107.1 | - | 3 | 437 | |||

Slc25a54
Genomic Map