siRNA Details
| Gene Symbol: Ttll9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083618.1 | NP_001077087.1 | 12 | 1081 | ||
| NM_029064.2 | NP_083340.2 | 12 | 1081 | |||
| GenBank mRNA | AK006027.1 | - | 12 | 1077 | ||
| AK015388.1 | - | 12 | 1060 | |||
| AK015740.1 | - | 12 | 1061 | |||
| AM690753.1 | - | 11 | 1025 | |||
| BC130024.1 | - | 12 | 1120 | |||
| BC148285.2 | - | 10 | 835 | |||

Ttll9
Genomic Map